Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triphenylphosphine

Catalog No. T8440925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
500 g
1 kg
10 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T8440925G 25 g
T84409500G 500 g
T84409100G 100 g
T8440910KG 10 kg
T844091G 1 g
T844091KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. T8440925G Supplier Sigma Aldrich Supplier No. T8440925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triphenylphosphine

Specifications

CAS 603-35-0
Melting Point 79°C to 81°C
Boiling Point 377°C
Molecular Formula C18H15P
Linear Formula (C6H5)3P
MDL Number MFCD00003043
Quantity 25 g
Synonym Phosphorustriphenyl
Molecular Weight (g/mol) 262.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.