Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine, 98%, Spectrum™ Chemical
SDP

Catalog No. 18614572 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
50 lb.
220 lb.
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-572 50 lb. Amber Glass Bottle
18-614-568 25 g Amber Glass Bottle
18-614-570 250 g Amber Glass Bottle
18-614-571 220 lb. Amber Glass Bottle
18-614-567 1 kg Amber Glass Bottle
18-614-569 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-614-572 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T112250LBBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 603-35-0

Specifications

CAS Max % 1
Boiling Point 360°C
Packaging Amber Glass Bottle
Quantity 50 lb.
Percent Purity 0.98
Grade Reagent
Physical Form Crystals or Flakes

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.