Learn More
Strem, An Ascensus Company CAS# 603-35-0. 500g. Triphenylphosphine, 99%. MFCD00003043
SDP
Catalog No. 5090110464
Encompass
CAS# 603-35-0. 500g. Triphenylphosphine, 99%. MFCD00003043. Molecular Weight: 262.28. Molecular Formula: (C6H5)3P. Color/form: white xtl. Strem# 15-6700. http://www.strem.com/catalog/v/15-6700/
Catalog No. 50-901-10464 Supplier Strem, An Ascensus Company Supplier No. 156700500G
CAS# 603-35-0. 500g. Triphenylphosphine, 99%. MFCD00003043. Molecular Weight: 262.28. Molecular Formula: (C6H5)3P. Color/form: white xtl. Strem# 15-6700. http://www.strem.com/catalog/v/15-6700/