Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Triphenylphosphine)Copper Hydride Hexamer

Catalog No. 3649755G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-497-55G 5 g
36-497-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 36-497-55G Supplier Sigma Aldrich Supplier No. 3649755G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triphenylgermanium hydride is a hydrogen donor.Some useful synthetic applications of (Triphenylphosphine)copperhydride hexamer include stoichiometric and catalytic hydrogenations, chemoselective conjugate reductions, as well as regiospecific and stereoselectiveconjugate hydride reductions.

Specifications

CAS 33636-93-0
Molecular Formula C108H96Cu6P6
Linear Formula [(C6H5)3PCuH]6
MDL Number MFCD00221518
Quantity 5 g
Synonym Cuprous hydride triphenylphosphine hexamer; Hydrido(triphenylphosphine)copper(I) hexamer; Stryker's reagent; Triphenylphosphine-Copper(I) hydride Hexamer
Molecular Weight (g/mol) 1961.04 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.