Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine oxide, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015554 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5554 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5554 Supplier MilliporeSigma Supelco Supplier No. 08902100MG
Add to Cart
Add to Cart

Specifications

CAS 791-28-6
Flash Point 180°C - Closed Cup
Linear Formula (C6H5)3 PO
MDL Number MFCD00002080
Melting Point 150°C to 157°C
Synonym Ph3 PO; TPPO; Triphenyl phosphorus oxide; Triphenylphosphine monoxide
Type Analytical Standard
Quantity 100 mg
Formula Weight 278.28
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.