Learn More
Chem-Impex International, Inc. Triphenylphosphine resin (0.9 - 1.6 meq/g, 200 - 400 mesh) | MFCD20489348 | 25G
Catalog No. 50500080
Encompass_Preferred
Triphenylphosphine resin (0.9 - 1.6 meq/g, 200 - 400 mesh), MFCD20489348, 25G
Catalog No. 50-500-080 Supplier Chem-Impex International, Inc. Supplier No. 0481425G
Triphenylphosphine resin (0.9 - 1.6 meq/g, 200 - 400 mesh), MFCD20489348, 25G