Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine sulfide, 99+%, Thermo Scientific™

Catalog No. AC159031000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC159031000 100 g
AC159030250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159031000 Supplier Thermo Scientific Chemicals Supplier No. 159031000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3878-45-3
Molecular Formula C18H15PS
Molecular Weight (g/mol) 294.35
MDL Number MFCD00004917
InChI Key VYNGFCUGSYEOOZ-UHFFFAOYSA-N
Synonym triphenylphosphine sulfide, phosphine sulfide, triphenyl, triphenylphosphine sulphide, triphenylphosphine monosulfide, triphenylphosphane sulfide, triphenylphosphino-1-thione, triphenylthioxophosphorane, triphenylphosphorus sulfide, wln: spr&r&r, triphenyl-, e5-phosphanethione
PubChem CID 19758
IUPAC Name triphenyl-λ⁵-phosphanethione
SMILES S=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 162.0°C to 164.0°C
Linear Formula (C6H5)3P(S)
Quantity 100 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: slightly soluble in organic solvents
Formula Weight 294.34
Chemical Name or Material Triphenylphosphine sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.