Learn More
Medchemexpress LLC Triphenylphosphonium-derivatized anethole dithiolethione (mitochondria-targeting H2S donor) | 1429061-80-2 | 96.4% | 721.77 g/mol | C37H38BrO2PS3 | 25MG
SDP
Catalog No. 5000235014
Encompass_Preferred
AP39 is a mitochondria-targeted hydrogen sulfide (H2S) donor used in biomedical research to elevate intracellular H2S and provide cytoprotective effects under oxidative stress. It is a triphenylphosphonium-derivatized anethole dithiolethione reported to preserve mitochondrial DNA integrity and to protect against myocardial reperfusion injury in preclinical models, with potential applications in neurodegenerative disease research.
  • Targets mitochondria to deliver hydrogen sulfide intracellularly.
  • Provides cytoprotective effects under oxidative stress.
  • Reported to preserve mitochondrial DNA integrity.
  • Demonstrated efficacy in preclinical myocardial reperfusion models.
  • Available as a solid in multiple milligram pack sizes for research use.
Catalog No. 50-002-35014 Supplier Medchemexpress LLC Supplier No. HY12612425MG
AP39 is a mitochondria-targeted hydrogen sulfide (H2S) donor used in biomedical research to elevate intracellular H2S and provide cytoprotective effects under oxidative stress. It is a triphenylphosphonium-derivatized anethole dithiolethione reported to preserve mitochondrial DNA integrity and to protect against myocardial reperfusion injury in preclinical models, with potential applications in neurodegenerative disease research.
  • Targets mitochondria to deliver hydrogen sulfide intracellularly.
  • Provides cytoprotective effects under oxidative stress.
  • Reported to preserve mitochondrial DNA integrity.
  • Demonstrated efficacy in preclinical myocardial reperfusion models.
  • Available as a solid in multiple milligram pack sizes for research use.