Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Triphenylphosphoranylidene)ketene, TCI America™
SDP

Catalog No. T25651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25651G Supplier TCI America Supplier No. T25651G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15596-07-3
Molecular Formula C20H15OP
Molecular Weight (g/mol) 302.313
MDL Number MFCD00040613
InChI Key MNASRBWCHRURHY-UHFFFAOYSA-N
Synonym Ketenylidene(triphenyl)phosphorane, Bestmann Ylide
PubChem CID 359581
IUPAC Name 2-(triphenyl-$l^{5}-phosphanylidene)ethenone
SMILES C1=CC=C(C=C1)P(=C=C=O)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 1 g
Formula Weight 302.31
Physical Form Crystalline Powder
Chemical Name or Material (Triphenylphosphoranylidene)ketene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.