Learn More
Strem, An Ascensus Company CAS# 789-25-3. 10g. Triphenylsilane, min. 97%. MFCD00003003
SDP
Catalog No. 5090114157
Encompass
CAS# 789-25-3. 10g. Triphenylsilane, min. 97%. MFCD00003003. Molecular Weight: 260.41. Molecular Formula: (C6H5)3SiH. Color/form: off-white pwdr. Strem# 93-1467. http://www.strem.com/catalog/v/93-1467/
Catalog No. 50-901-14157 Supplier Strem, An Ascensus Company Supplier No. 93146710G
CAS# 789-25-3. 10g. Triphenylsilane, min. 97%. MFCD00003003. Molecular Weight: 260.41. Molecular Formula: (C6H5)3SiH. Color/form: off-white pwdr. Strem# 93-1467. http://www.strem.com/catalog/v/93-1467/