Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triphenylsulfonium-d5 chloride | 99.6% | 303.86 g·mol⁻¹ | C18H10D5ClS | 1 MG
SDP

Catalog No. 5000283574
Encompass_Preferred

Triphenylsulfonium-d5 chloride is a deuterium-labeled reagent used as an isotope-labeled analogue of triphenylsulfonium chloride for analytical and research applications. Supplied as a white to off-white solid with reported high purity, it is commonly used as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High reported purity (≈99.6%).
  • Molecular formula C18H10D5ClS; molecular weight 303.86 g·mol⁻¹.
  • Available in small pack sizes suitable for analytical use, e.g., 1 mg.
  • White to off-white solid with documented analytical data (HNMR, RP-HPLC, MS, COA).
  • Recommended storage: 4°C sealed, protect from moisture; in solution store at -80°C for long-term stability.
  • Intended for research use only; not for human or animal therapeutic use.

Catalog No. 50-002-83574 Supplier Medchemexpress LLC Supplier No. HYW140598S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triphenylsulfonium-d5 chloride is a deuterium-labeled reagent used as an isotope-labeled analogue of triphenylsulfonium chloride for analytical and research applications. Supplied as a white to off-white solid with reported high purity, it is commonly used as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High reported purity (≈99.6%).
  • Molecular formula C18H10D5ClS; molecular weight 303.86 g·mol⁻¹.
  • Available in small pack sizes suitable for analytical use, e.g., 1 mg.
  • White to off-white solid with documented analytical data (HNMR, RP-HPLC, MS, COA).
  • Recommended storage: 4°C sealed, protect from moisture; in solution store at -80°C for long-term stability.
  • Intended for research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.