Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylsulfonium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. T16081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16081G 1 g
T16085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16081G Supplier TCI America Supplier No. T16081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 437-13-8
Molecular Formula C18H15BF4S
Molecular Weight (g/mol) 350.18
MDL Number MFCD00671581
InChI Key RTWMEMGVYYTCOZ-UHFFFAOYSA-N
PubChem CID 2760877
IUPAC Name tetrafluoroboranuide; triphenylsulfanium
SMILES F[B-](F)(F)F.C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 194°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 350.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylsulfonium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.