Learn More
AdipoGen Triphenyltetrazolium chloride, 298-96-4, MFCD00011963, 100 g
SDP
Catalog No. 50596124
Encompass_Preferred
MF: C19H15ClN4, MW: 334.8, ≥99%, Appearance: Off-white to yellow powder. Triphenyl tetrazolium chloride, TTC, or simply tetrazolium chloride is a redox indicator commonly used in biochemical experiments especially to indicate cellular respiration.
Catalog No. 50-596-124 Supplier AdipoGen Supplier No. CDXT0026G100
MF: C19H15ClN4, MW: 334.8, ≥99%, Appearance: Off-white to yellow powder. Triphenyl tetrazolium chloride, TTC, or simply tetrazolium chloride is a redox indicator commonly used in biochemical experiments especially to indicate cellular respiration.