Learn More
AdipoGen Triphenyltetrazolium chloride, 298-96-4, MFCD00011963, 25 g
SDP
Catalog No. 50596123
Encompass_Preferred
MF: C19H15ClN4, MW: 334.8, ≥99%, Appearance: Off-white to yellow powder. Triphenyl tetrazolium chloride, TTC, or simply tetrazolium chloride is a redox indicator commonly used in biochemical experiments especially to indicate cellular respiration.
Catalog No. 50-596-123 Supplier AdipoGen Supplier No. CDXT0026G025
MF: C19H15ClN4, MW: 334.8, ≥99%, Appearance: Off-white to yellow powder. Triphenyl tetrazolium chloride, TTC, or simply tetrazolium chloride is a redox indicator commonly used in biochemical experiments especially to indicate cellular respiration.