Learn More
Crescent Chemical Co Inc TRIPHENYLTETRAZOLIUM CHLORIDE
SDP
Catalog No. 50248224
Encompass_Preferred
Triphenyltetrazolium chloride analytical grade For microbiology and histochemistry. Vital staining of cells. Size - 10G Storage Conditions - +15 °C TO +30 °C Catalog Number - 37130.03 CAS 298-96-4
Catalog No. 50-248-224 Supplier Crescent Chemical Co Inc Supplier No. SE37130.03
Triphenyltetrazolium chloride analytical grade For microbiology and histochemistry. Vital staining of cells. Size - 10G Storage Conditions - +15 °C TO +30 °C Catalog Number - 37130.03 CAS 298-96-4