Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyltin acetate, 97%, Thermo Scientific™

Catalog No. AA4578909
Change view
Click to view available options
Quantity:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4578909 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4578909 Supplier Thermo Scientific Chemicals Supplier No. 4578909

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 900-95-8
Molecular Formula C20H18O2Sn
Linear Formula (C6H5)3SnOOCCH3
Quantity 10g
MDL Number MFCD00014977
UN Number UN3146
Beilstein 4146107
Merck Index 14,9745
Synonym fentin acetate, triphenyltin acetate, phentin acetate, phentinoacetate, batasan, brestan, acetoxytriphenylstannane, fentinacetat, lirostanol, liromatin
InChI Key WDQNIWFZKXZFAY-UHFFFAOYSA-M
SMILES CC(=O)O[Sn](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
IUPAC Name triphenylstannyl acetate
Molecular Weight (g/mol) 409.072
PubChem CID 16682804
ChEBI CHEBI:81918
Formula Weight 409.05
Percent Purity 97%
Chemical Name or Material Triphenyltin acetate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H311-H330-H372-H351-H361-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
RTECSNumber WH6650000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.