Learn More
Medchemexpress LLC Triphenyltin chloride-d15 | 358731-94-9 | MFCD01861652 | 99.7% | 400.57 g·mol⁻1 | C18D15ClSn | 10 MG
SDP
Catalog No. 5000283399
Encompass_Preferred
Triphenyltin chloride-d15 is the deuterium-labeled analogue of triphenyltin chloride supplied as a high-purity reference standard for research and analytical use. It is intended for use as an internal standard in mass spectrometry, isotopic tracing, and labeling studies, and is provided in small, laboratory-scale quantities.
  • Deuterium-labeled (D15) isotopologue for accurate mass spectrometry quantitation.
  • High purity suitable for reference standard and analytical use.
  • Small-scale quantities for laboratory research and method development.
  • Useful for isotopic tracing and labeling experiments.
  • Provided as a solid with defined molecular weight and CAS registry number.
Catalog No. 50-002-83399 Supplier Medchemexpress LLC Supplier No. HYW134259S10MG
Triphenyltin chloride-d15 is the deuterium-labeled analogue of triphenyltin chloride supplied as a high-purity reference standard for research and analytical use. It is intended for use as an internal standard in mass spectrometry, isotopic tracing, and labeling studies, and is provided in small, laboratory-scale quantities.
  • Deuterium-labeled (D15) isotopologue for accurate mass spectrometry quantitation.
  • High purity suitable for reference standard and analytical use.
  • Small-scale quantities for laboratory research and method development.
  • Useful for isotopic tracing and labeling experiments.
  • Provided as a solid with defined molecular weight and CAS registry number.