Learn More
Medchemexpress LLC Triphenyltin chloride-d15 | 358731-94-9 | MFCD01861652 | 99.7% | 400.57 g/mol | C18D15ClSn | 5 MG
SDP
Catalog No. 5000283264
Encompass_Preferred
Triphenyltin chloride-d15 is the deuterium-labeled analogue of triphenyltin chloride used as a stable isotope reference standard and research reagent for analytical chemistry, mass spectrometry, and isotopic labeling studies. It is supplied as a powder and characterized by high isotopic enrichment and chemical purity.
  • Deuterium-labeled standard for analytical and mass spectrometry applications.
  • High chemical purity and isotopic enrichment for accurate quantitation.
  • Supplied as a stable powder with defined storage conditions.
  • Useful as an internal standard and reference material in method development.
  • Characterized by molecular formula C18D15ClSn and molecular weight 400.57 g/mol.
Catalog No. 50-002-83264 Supplier Medchemexpress LLC Supplier No. HYW134259S5MG
Triphenyltin chloride-d15 is the deuterium-labeled analogue of triphenyltin chloride used as a stable isotope reference standard and research reagent for analytical chemistry, mass spectrometry, and isotopic labeling studies. It is supplied as a powder and characterized by high isotopic enrichment and chemical purity.
  • Deuterium-labeled standard for analytical and mass spectrometry applications.
  • High chemical purity and isotopic enrichment for accurate quantitation.
  • Supplied as a stable powder with defined storage conditions.
  • Useful as an internal standard and reference material in method development.
  • Characterized by molecular formula C18D15ClSn and molecular weight 400.57 g/mol.