Learn More
Strem, An Ascensus Company CAS# 379-52-2. 5g. Triphenyltin fluoride, min. 97%. MFCD00015356
SDP
Supplier:  Strem, An Ascensus Company 9350615G
Encompass
CAS# 379-52-2. 5g. Triphenyltin fluoride, min. 97%. MFCD00015356. Molecular Weight: 368.99. Molecular Formula: (C6H5)3SnF. Color/form: white pwdr. Strem# 93-5061. http://www.strem.com/catalog/v/93-5061/
Catalog No. 50-901-15614