Learn More
Bellco Glass, Inc. Triple Addition Assembly ,45mm Universal
SDP
Supplier:  Bellco Glass, Inc. 196403250
Encompass_Preferred
Triple Addition Assembly. For GL45 sidearms. Manufactured from 316l stainless steel with electropolish finish ra 32 or better inside and out. Supply with PP blue Open top s/cap & white silicone liner. All tubes: 1/4" OD x 4 1/4" long. Inseration Length: 3 3/32". Application: Individual ports for liquid additions or redundant vent/overlay. Vertical Side Arm vessels for acid, base, antifoam, media & other additions. MADE IN THE USA
Catalog No. 50-194-4738