Learn More
Med Vet International First Aid Antibiotic Water Jel Ointment 0.9 Gram Individual Packet, WJTA1728, 144/Box
Catalog No. 503043867
Encompass_Preferred
Each gram contains 400 IU bacitracin, 3.5 milligrams neomycin and 5,000 IU of polymyxin B Comparable to Neosporin Original Ointment Ingredients help prevent infection in minor cuts, scrapes and burns Packaged in a 0.9-gram dose packet for individual application without cross-contamination Neosporin is a registered trademark of Johnson & Johnson
Catalog No. 50-304-3867 Supplier Med Vet International Supplier No. 943299BX
Each gram contains 400 IU bacitracin, 3.5 milligrams neomycin and 5,000 IU of polymyxin B Comparable to Neosporin Original Ointment Ingredients help prevent infection in minor cuts, scrapes and burns Packaged in a 0.9-gram dose packet for individual application without cross-contamination Neosporin is a registered trademark of Johnson & Johnson