Learn More
Med Vet International First Aid Antibiotic Medi-First Ointment 0.5 Gram Individual Packet, 22373, 25/Box
Catalog No. 503044690
Encompass_Preferred
For effective protection against infection from minor cuts, burns and abrasions Active ingredients include 400 IU bacitracin, 5 mg neomycin and 5,000 IU polymyxin B (similar to Neosporin – a registered trademark of Johnson & Johnson) First aid ointment is packaged in half-gram packets, 25 packets per box for easy distribution as part of workplace first aid kit or as a "just in case" item in your travel toiletries bag
Catalog No. 50-304-4690 Supplier Med Vet International Supplier No. 337028BX
For effective protection against infection from minor cuts, burns and abrasions Active ingredients include 400 IU bacitracin, 5 mg neomycin and 5,000 IU polymyxin B (similar to Neosporin – a registered trademark of Johnson & Johnson) First aid ointment is packaged in half-gram packets, 25 packets per box for easy distribution as part of workplace first aid kit or as a "just in case" item in your travel toiletries bag