Learn More
Med Vet International First Aid Antibiotic Water Jel Ointment 0.9 Gram Individual Packet, WJTA1800.00.000, 1800/Case
Catalog No. 503043626
Encompass_Preferred
400 IU bacitracin; 3.5 mg neomycin; and 5,000 IU polymyxin B per dose First aid ointment prevents infection of cuts and other injuries
Catalog No. 50-304-3626 Supplier Med Vet International Supplier No. 769307CS
400 IU bacitracin; 3.5 mg neomycin; and 5,000 IU polymyxin B per dose First aid ointment prevents infection of cuts and other injuries