Learn More
Grainger Adam Equipment Triple Beam Balance Legs, Cast Aluminum, Black, 1 ea.
Catalog No. 502795931
Encompass_Preferred
These tripod legs are replacement parts designed to support the repair and maintenance of TBB triple beam balances. These legs are engineered to raise the balance height, which facilitates density and specific gravity calculations and enables the measurement of bulkier samples via below-balance weighing.
  • Three-point tripod design ensures inherent stability on uneven floors and ground
  • Constructed from durable cast aluminum with a smooth finish
  • Compatible with specific triple beam balance models
  • Designed for functional support and height adjustment
Catalog No. 50-279-5931 Supplier Grainger Supplier No. 19YP04
These tripod legs are replacement parts designed to support the repair and maintenance of TBB triple beam balances. These legs are engineered to raise the balance height, which facilitates density and specific gravity calculations and enables the measurement of bulkier samples via below-balance weighing.
  • Three-point tripod design ensures inherent stability on uneven floors and ground
  • Constructed from durable cast aluminum with a smooth finish
  • Compatible with specific triple beam balance models
  • Designed for functional support and height adjustment