Learn More
CHALEX LLC TRIPLE-CHAMBER MCMASTER SLIDE
Catalog No. NC2165357
Encompass
Triple-Chamber McMaster SlideSlide size:1-7/16″ x 3″ (36 mm x 75 mm)Grid size:1 cm x 2 cm (1 each over separate chambers)Grid Style/Color:Green PrintedVolume under grid:0.30 mlMaterial:all polycarbonate
This item is not returnable. View return policy
Catalog No. NC2165357 Supplier CHALEX LLC Supplier No. 3CHAMBER POLY
Triple-Chamber McMaster SlideSlide size:1-7/16″ x 3″ (36 mm x 75 mm)Grid size:1 cm x 2 cm (1 each over separate chambers)Grid Style/Color:Green PrintedVolume under grid:0.30 mlMaterial:all polycarbonate