Learn More
MP Biomedicals™ Triple Color Precision Protein Ladder (10-180 kDa)
Shop All MP Biomedicals, Inc ProductsDescription
MP Biomedicals Triple Color Precision Protein Ladder (10-180 kDa) is a ready-to-use three-color protein standard with 10 pre-stained proteins covering a wide range of molecular weights from 10 to 180 kDa. This marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes, and approximating the size of proteins.
Specifications
Specifications
| Concentration | 0.1 to 0.4 mg/mL of each protein in the buffer |
| Formulation | In 20 mM Tris-Phosphate buffer (pH 7.5) containing 2% SDS, 0.2 mM DTT, 3.6 M urea, and 15% (v/v) glycerol |
| Quantity | 50 μL |
| Regulatory Status | RUO |
| Content And Storage | -20°C |
| Solubility | Ready to use without dilutions |
Safety and Handling
For research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.