Learn More
Electron Microscopy Sciences GLACIER™ Medium Ice Pan 4 Litre Triple, 12.25" x 8.75" x 4.75"
SDP
Catalog No. 501899111
Encompass_Preferred
The ice pans are made of triple density expanded polystyrene creating a durable container that is virtually unbreakable. Thick pan walls do not sweat or leave puddles of condensation on the benchtop. It preserves ice up to 30% longer than conventional ice pans.
Extremely lightweight and with molded lip around the edge are easy to handle An inner edge enables the pans to stack on top of one another for easy storage and to minimize bench top space.
Resistant to mild chemicals as well as ice-alcohol slurry, dry ice, and liquid nitrogen. Not recommended for long-term exposure to harsh chemicals or corrosives.
Capacity: 4 L
Dimensions: 12.25" x 8.75" x 4.75"
Catalog No. 50-189-9111 Supplier Electron Microscopy Sciences Supplier No. 633111M
The ice pans are made of triple density expanded polystyrene creating a durable container that is virtually unbreakable. Thick pan walls do not sweat or leave puddles of condensation on the benchtop. It preserves ice up to 30% longer than conventional ice pans.
Extremely lightweight and with molded lip around the edge are easy to handle An inner edge enables the pans to stack on top of one another for easy storage and to minimize bench top space.
Resistant to mild chemicals as well as ice-alcohol slurry, dry ice, and liquid nitrogen. Not recommended for long-term exposure to harsh chemicals or corrosives.
Capacity: 4 L
Dimensions: 12.25" x 8.75" x 4.75"