Learn More
Rockland Immunochemicals Triple FLAG Peptide - 000-000-H47
SDP
Supplier:  Rockland Immunochemicals 000000H47
Encompass
The Triple FLAG Peptide is a synthetic peptide of 23 amino acid residues (molecular weight of 2861.9 daltons). The Asp-Tyr-Lys-Xaa-Xaa-Asp motif is repeated three times in the peptide. Eight amino acids at the C-terminus make up the classic FLAG sequence (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys or DYKDDDDK). The triple FLAG peptide is intended for immunoaffinity chromatography to allow elution under non-denaturing conditions by competitive elution with excess of free triple FLAG Peptide.
Catalog No. 50-105-7197