Learn More
Euthanex Integrated Imaging Triple Breather Block
Catalog No. 501540154
Encompass_Preferred
Integrated Imaging Triple Breather Block can be permanentely integrated into a wide range of imaging systems. The low profile, all plastic construction breathers include threaded mounting holes for simple integration. Designed for use with animals weighing up to 500 grams. Includes all tubing and connectors.
Catalog No. 50-154-0154 Supplier Euthanex Supplier No. EZ109013
Integrated Imaging Triple Breather Block can be permanentely integrated into a wide range of imaging systems. The low profile, all plastic construction breathers include threaded mounting holes for simple integration. Designed for use with animals weighing up to 500 grams. Includes all tubing and connectors.