Learn More
Chemglass Life Sciences Triple Insulated Hose, 4ft Length
SDP
Catalog No. 50872377
Encompass_Preferred
Triple Insulated Hose, 4ft Length. For connection of circulators to Chemglass reactors. Hoses are flexible stainless steel tubing, insulated with fiberglass and silicone. Temperature range -80 to 200° C. Both ends have a M16 x 1 Female fitting for attachment directly to our inlet/outlet connections see CG-1969-A. Adapters are available for connection to most circulators.PLEASE NOTE: TWO HOSES ARE REQUIRED PER REACTOR.
Catalog No. 50-872-377 Supplier Chemglass Life Sciences Supplier No. CG1969B04
Triple Insulated Hose, 4ft Length. For connection of circulators to Chemglass reactors. Hoses are flexible stainless steel tubing, insulated with fiberglass and silicone. Temperature range -80 to 200° C. Both ends have a M16 x 1 Female fitting for attachment directly to our inlet/outlet connections see CG-1969-A. Adapters are available for connection to most circulators.PLEASE NOTE: TWO HOSES ARE REQUIRED PER REACTOR.