Learn More
Chemglass Life Sciences Triple Port Adapter, M10 Thread, 1/8"OD Tubing, 316L Stainless Steel, Electro Polished Ra20
SDP
Catalog No. NC2151478
Encompass
316L stainless steel triple port assembly fits into a 10mm head port plate.The assembly incorporates three separate pipes into on assembly for efficient use of space.Each port can be used individually for reagent addition.M10 threaded adapter has (3) 1/8 OD tubing ports for connection to flexible tubing.Inside the reactor the tubes are 7mm long; above the headplate they are 1.59, 2.09, and 2.59long.Electropolished to Ra20 or better.
This item is not returnable. View return policy
Catalog No. NC2151478 Supplier Chemglass Life Sciences Supplier No. CLS1380323
316L stainless steel triple port assembly fits into a 10mm head port plate.The assembly incorporates three separate pipes into on assembly for efficient use of space.Each port can be used individually for reagent addition.M10 threaded adapter has (3) 1/8 OD tubing ports for connection to flexible tubing.Inside the reactor the tubes are 7mm long; above the headplate they are 1.59, 2.09, and 2.59long.Electropolished to Ra20 or better.