Learn More
Chemglass Life Sciences Triple Port Adapter, M18, for Bio Reactors
SDP
Catalog No. 5000980590
Encompass
The triple inlet assembly fits into an 18mm head plate port. The assembly incorporates three separate pipes into one assembly for efficient use of space. Each port can be used indivually for reagent addition. M18 threaded adapter has (3) 1/8" (3. 2mm) tubing ports for connection to flexible tubing. Inside the reactor, the 1/8" tubes are 3/4", 1" and 1-1/8" long. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-009-80590 Supplier Chemglass Life Sciences Supplier No. CLS138009
The triple inlet assembly fits into an 18mm head plate port. The assembly incorporates three separate pipes into one assembly for efficient use of space. Each port can be used indivually for reagent addition. M18 threaded adapter has (3) 1/8" (3. 2mm) tubing ports for connection to flexible tubing. Inside the reactor, the 1/8" tubes are 3/4", 1" and 1-1/8" long. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.