Learn More
Alkali Scientific ASI™ Triple-Pure Starter Kit, High impact Zirconium Beads, includes 10 each of 0.1, 0.5, 1.0, 1.5 and 3.0mm
SDP
Catalog No. 503027506
Encompass_Preferred
These Triple-Pure Starter Kit, High impact Zirconium Beads, and come with standard acid-washed glass beads, triple-pure zirconium beads, or stainless steel beads. The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease. The stainless steel beads are acid-washed as well. All tubes are non-skirted with screw-cap. Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.
Catalog No. 50-302-7506 Supplier Alkali Scientific Supplier No. RS70129
These Triple-Pure Starter Kit, High impact Zirconium Beads, and come with standard acid-washed glass beads, triple-pure zirconium beads, or stainless steel beads. The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease. The stainless steel beads are acid-washed as well. All tubes are non-skirted with screw-cap. Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.