Learn More
Mopec Triple Scrub Station
SDP
Catalog No. 22046216 Shop All Mopec Products
Change view
Click to view available options
Certifications/Compliance:
ADA Compliant
Non - ADA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
22-046-216 Non - ADA
22-046-217 ADA Compliant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-046-216 Supplier Mopec Supplier No. SS103
Add to Cart
Add to Cart
ADA-compliant model available, our Scrub Stations ensure inclusivity without compromising performance, laboratories prioritizing safety.
  • Thermostatically-controlled mixing faucet with gooseneck spout
  • Backflow protection for all plumbing fixtures to prevent backflow of contaminated water
  • Rapid and positive drainage
  • Access panel to get to plumbing
  • ADA wheelchair accessible: (SS101-ADA)
  • OA-151: Hot and cold foot valve
Specifications
Certifications/Compliance Non - ADA
Color Silver
Material Stainless Steel (304)
Type Multistation Hand Sink