Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Triple Sugar Iron Agar, Prepared Media Tubes
SDPSCI_ED

Catalog No. S42842 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S42842 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S42842 Supplier Carolina™ Supplier No. 827442
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Carefully tested for sterility and ability to support the characteristic growth of appropriate organisms.

  • Sterile
  • Ready-to-use media
  • No autoclave needed
  • Slant agar tubes
Note: C of A unavailable for this item.

Specifications

Content And Storage 2°C to 8°C
Description Sterile
Manufacturing Origin US with Globally Sourced Materials
Media Type Triple sugar iron slant agar medium
Product Type Agar Slants
Sterility Sterile
Target Organism Class Enteric Bacteria
Test Type Microbial Differentiation
For Use With (Application) Culture of organisms
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.