Learn More
Alkali Scientific Triple Sugar Iron Agar Slant 4mL 13 x 100 mm S/CAP GL, 10/Pk
SDP
Catalog No. 5000662803
Encompass_Preferred
Triple Sugar Iron Agar Slant 4mL 13 x 100mm S/CAP GL, 10/Pk. A differential medium used in laboratories for the identification of microorganisms based on sugar fermentation and hydrogen sulfide production. The slant format provides an optimal surface for microbial growth. This 4mL slant is ideal for laboratories that require compact yet effective testing environments.
Catalog No. 50-006-62803 Supplier Alkali Scientific Supplier No. TSIT00
Triple Sugar Iron Agar Slant 4mL 13 x 100mm S/CAP GL, 10/Pk. A differential medium used in laboratories for the identification of microorganisms based on sugar fermentation and hydrogen sulfide production. The slant format provides an optimal surface for microbial growth. This 4mL slant is ideal for laboratories that require compact yet effective testing environments.