Learn More
Alkali Scientific Triple Sugar Iron Agar Slant, 9mL, 16x125mm Screw Cap, 10/Pk
SDP
Catalog No. 5000663273
Encompass_Preferred
Triple Sugar Iron (TSI) Agar Slant is a differential medium formulated to identify and characterize enteric bacteria based on their ability to ferment glucose, lactose, and sucrose, as well as produce hydrogen sulfide and gas. Commonly used in clinical, pharmaceutical, and industrial microbiology laboratories, TSI Agar supports reliable biochemical differentiation of gram negative enteric organisms. Each 9mL slant is prepared in a 16x125mm screw cap glass tube to maintain sterility during storage, transport, and incubation. The medium enables detection of carbohydrate fermentation patterns and hydrogen sulfide production, supporting accurate identification of Enterobacteriaceae and related organisms in diagnostic workflows and routine microbiological testing.
Catalog No. 50-006-63273 Supplier Alkali Scientific Supplier No. TSIAT00
Triple Sugar Iron (TSI) Agar Slant is a differential medium formulated to identify and characterize enteric bacteria based on their ability to ferment glucose, lactose, and sucrose, as well as produce hydrogen sulfide and gas. Commonly used in clinical, pharmaceutical, and industrial microbiology laboratories, TSI Agar supports reliable biochemical differentiation of gram negative enteric organisms. Each 9mL slant is prepared in a 16x125mm screw cap glass tube to maintain sterility during storage, transport, and incubation. The medium enables detection of carbohydrate fermentation patterns and hydrogen sulfide production, supporting accurate identification of Enterobacteriaceae and related organisms in diagnostic workflows and routine microbiological testing.