Learn More
Bioworld Triple Sugar Iron (TSI) Agar, 2 kg
Catalog No. 50254382
Encompass_Preferred
Triple Sugar Iron (TSI) AgarFor the differentiation of gram-negative enteric bacilli based on carbohydrate fermentation and the production of hydrogen sulfidepH: 6.9-7.1Working Concentration: 60 g/LDehydrated Appearance: pink, homogeneous powderPrepared Appearance: orange to orange-red solutionSoluble in boiling waterContains:Pancreatic Digest of Casein Peptic Digest of Animal TissueYeast Enriched PeptoneDextrose Lactose Sucrose Ferrous Ammonium Sulfate Sodium Chloride Sodium Thiosulfate Phenol Red Agar
Catalog No. 50-254-382 Supplier Bioworld Supplier No. 306200853
Triple Sugar Iron (TSI) AgarFor the differentiation of gram-negative enteric bacilli based on carbohydrate fermentation and the production of hydrogen sulfidepH: 6.9-7.1Working Concentration: 60 g/LDehydrated Appearance: pink, homogeneous powderPrepared Appearance: orange to orange-red solutionSoluble in boiling waterContains:Pancreatic Digest of Casein Peptic Digest of Animal TissueYeast Enriched PeptoneDextrose Lactose Sucrose Ferrous Ammonium Sulfate Sodium Chloride Sodium Thiosulfate Phenol Red Agar