Learn More
Omnimed Inc Triple Wire Glove Box Holder/Dispenser
SDP
Catalog No. NC3956478
Encompass_Preferred
Omnimed Wire Glove Box Holders are designed for durability and ease of use, making them perfect for medical, laboratory, food preparation, or any environment where protective gloves are essential and cross-contamination must be minimized. Constructed from heavy-gauge steel wire and coated with a durable, easy-to-clean white powder finish, these glove box holders can accommodate up to three glove boxes. The simple design allows for easy loading and access, and the units can be mounted either vertically or horizontally to suit your space and workflow needs.
Catalog No. NC3956478 Supplier Omnimed Inc Supplier No. 305375
Omnimed Wire Glove Box Holders are designed for durability and ease of use, making them perfect for medical, laboratory, food preparation, or any environment where protective gloves are essential and cross-contamination must be minimized. Constructed from heavy-gauge steel wire and coated with a durable, easy-to-clean white powder finish, these glove box holders can accommodate up to three glove boxes. The simple design allows for easy loading and access, and the units can be mounted either vertically or horizontally to suit your space and workflow needs.