Learn More
Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar with VHP Indicator and 2D Barcode
Catalog No. R111TW3 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
85 mm plate, standard lid
Catalog No. Format
R111TW3 85 mm plate, standard lid
1 options
This item is not returnable. View return policy
Catalog No. R111TW3 Supplier Thermo Scientific™ Supplier No. R111TW3
Add to Cart
Add to Cart
This item is not returnable. View return policy
Carry out environmental monitoring in your most regulated environments using Triple Wrapped Plates, for passive and active air sampling and surface testing applications.
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator and 2D Barcode.
Choose from standard or locking lid formats to meet your environmental monitoring workflow needs. The standard lid provides a familiar format for routine environmental monitoring, while the locking lid provides a secure closure option for workflows where additional plate security is preferred.
Achieve confidence in your results and ensure optimum media growth assurance using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.
Three protective layers reduce risk of contamination:
  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover
  • Conveniently sold in packs of 100 which are sealed for security
The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY
Specifications
Color Ivory, transparent
Format 85 mm plate, standard lid
Product Type Microbiology Media
Fill Weight 30 grams (+/- 0.5)
Packaging Type No cardboard inner packaging, triple wrapped-irradiated, with VHP barrier wrap
Quantity 100/pk