Learn More
Ecolab Ecolab Klerwipe™ 70/30 IPA Wipes, Sterile, Gamma Irradiated, Triple-Bagged Packaging, 69.5 Qty
Catalog No. NC2330085
Encompass
Ecolab Klerwipe™ 70/30 IPA Blended with WFI Sterile Pouch Wipes are premium, ready-to-use wipes designed for critical cleanroom and scientific applications. Engineered with 70% isopropyl alcohol solution and formulated with Water for Injection (WFI), they ensure consistent cleaning while minimizing contamination risk. Manufactured in a Grade C/ISO Class 5 cleanroom, these ultrasonically sealed wipes provide robust sterility and effective cleaning for aseptic processing and product contact surfaces.
Catalog No. NC2330085 Supplier Ecolab Supplier No. 6600072
Ecolab Klerwipe™ 70/30 IPA Blended with WFI Sterile Pouch Wipes are premium, ready-to-use wipes designed for critical cleanroom and scientific applications. Engineered with 70% isopropyl alcohol solution and formulated with Water for Injection (WFI), they ensure consistent cleaning while minimizing contamination risk. Manufactured in a Grade C/ISO Class 5 cleanroom, these ultrasonically sealed wipes provide robust sterility and effective cleaning for aseptic processing and product contact surfaces.