Learn More
Abcam TripleStain IHC Kit: M&M&R on Human tissue (DAB, AP/Red & HRP/Green), 60ML
Catalog No. 502590563
Encompass_Preferred
Abcam's TripleStain IHC kit (ab183286) is designed to use with user supplied two mouse primary antibodies and one rabbit primary antibody to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0563 Supplier Abcam Supplier No. AB18328660ML
Add to Cart
Add to Cart
Request a Quote
Abcam's TripleStain IHC kit (ab183286) is designed to use with user supplied two mouse primary antibodies and one rabbit primary antibody to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement