Learn More
Abcam TripleStain IHC Kit: M&M&R on Human tissue (DAB, AP/Red & HRP/Green), 18ML
Catalog No. 502590564
Encompass_Preferred
Abcam's TripleStain IHC kit (ab183286) is designed to use with user supplied two mouse primary antibodies and one rabbit primary antibody to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0564 Supplier Abcam Supplier No. AB18328618ML
Abcam's TripleStain IHC kit (ab183286) is designed to use with user supplied two mouse primary antibodies and one rabbit primary antibody to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement