Learn More
Abcam TripleStain IHC Kit: M&M&R on Human tissue (DAB, AP/Red & HRP/Green), 6ML
Catalog No. 502590562
Encompass_Preferred
Abcam's TripleStain IHC kit (ab183286) is designed to use with user supplied two mouse primary antibodies and one rabbit primary antibody to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0562 Supplier Abcam Supplier No. AB1832866ML
Abcam's TripleStain IHC kit (ab183286) is designed to use with user supplied two mouse primary antibodies and one rabbit primary antibody to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement