Learn More
Abcam TripleStain IHC Kit: M&R&G on human tissue (DAB, AP/Red & HRP/Green), 18ML
Catalog No. 502590558
Encompass_Preferred
Abcam's TripleStain IHC Kit (ab183290) is designed to be used with user-supplied one goat, one mouse and one rabbit primary antibodies to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0558 Supplier Abcam Supplier No. AB18329018ML
Abcam's TripleStain IHC Kit (ab183290) is designed to be used with user-supplied one goat, one mouse and one rabbit primary antibodies to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement