Learn More
Abcam TripleStain IHC Kit: M&R&G on human tissue (DAB, AP/Red & HRP/Green), 60ML
Catalog No. 502590557
Encompass_Preferred
Abcam's TripleStain IHC Kit (ab183290) is designed to be used with user-supplied one goat, one mouse and one rabbit primary antibodies to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0557 Supplier Abcam Supplier No. AB18329060ML
Add to Cart
Add to Cart
Request a Quote
Abcam's TripleStain IHC Kit (ab183290) is designed to be used with user-supplied one goat, one mouse and one rabbit primary antibodies to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement