Learn More
Abcam TripleStain IHC Kit: M&R&Rt on rodent tissue (DAB, HRP/Green & AP/Red), 6ML
Catalog No. 502590553
Encompass_Preferred
The TripleStain IHC kit (ab183297) is designed to be used with user supplied mouse, rat and rabbit primary antibodies to detect three distinct antigens on mouse tissue and cell samples or human tissue.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0553 Supplier Abcam Supplier No. AB1832976ML
The TripleStain IHC kit (ab183297) is designed to be used with user supplied mouse, rat and rabbit primary antibodies to detect three distinct antigens on mouse tissue and cell samples or human tissue.
The product is subject to the following: Abcam Restricted Use Statement