Learn More
Abcam TripleStain IHC Kit: M&R&G on human tissue (DAB, AP/Red & DAB/Ni), 18ML
Catalog No. 502677847
Encompass_Preferred
Abcam's TripleStain IHC Kit (ab183291) is designed to be used with user-supplied goat, mouse and rabbit primary antibodies to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-7847 Supplier Abcam Supplier No. AB18329118ML
Abcam's TripleStain IHC Kit (ab183291) is designed to be used with user-supplied goat, mouse and rabbit primary antibodies to detect three distinct antigens on a single human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement