Learn More
Abcam TripleStain IHC Kit: M&R&Rt on rodent tissue (DAB, DAB/Ni & AP/Red), 18ML
Catalog No. 502677844
Encompass_Preferred
The TripleStain IHC Kit (ab183298) is designed to be used with user-supplied mouse, rat and rabbit primary antibodies to detect three distinct antigens on mouse tissue and cell samples or human tissue.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-7844 Supplier Abcam Supplier No. AB18329818ML
The TripleStain IHC Kit (ab183298) is designed to be used with user-supplied mouse, rat and rabbit primary antibodies to detect three distinct antigens on mouse tissue and cell samples or human tissue.
The product is subject to the following: Abcam Restricted Use Statement