Learn More
Abcam TripleStain IHC Kit: R&R&M on human tissue (DAB, AP/Red & Green/HRP), 60ML
Catalog No. 502590560
Encompass_Preferred
Abcam's TripleStain IHC Kit (ab183288) is designed to be used with one mouse and two rabbit primary antibodies, supplied by the user, to detect three distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0560 Supplier Abcam Supplier No. AB18328860ML
Add to Cart
Add to Cart
Request a Quote
Abcam's TripleStain IHC Kit (ab183288) is designed to be used with one mouse and two rabbit primary antibodies, supplied by the user, to detect three distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement